Ammonium hexafluorotitanate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:16962-40-6
:
Formula:H₈F₆N₂Ti
MDL Number:MFCD00010888
:197.95 g/mol
ADR:6.1,III
:M-Clarity™ quality level = MQ100
EINECS:241-036-9
UN:3288
Frequently Bought Together