
Ammonium hexafluorophosphate 99%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16941-11-0
:
Formula:H₄F₆NP
MDL Number:MFCD00064642
:163 g/mol
Merck Index:12,00557
Melting Pt:198 °C (dec.)
Density:2.18 g/cm³
Storage Temperature:Ambient
EINECS:241-009-1
UN:3260
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White Crystalline powder
- Elemental Analysis:≥98.5 %
Specification Test Results
Frequently Bought Together