Tetrapropylammonium perruthenate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:114615-82-6
:
Formula:C₁₂H₂₈NO₄Ru
MDL Number:MFCD00074914
Melting Pt:165 °C
ADR:5.1,III
:M-Clarity™ quality level = MQ100
:351.43 g/mol
Storage Temperature:Refrigerator
UN:1479
Frequently Bought Together