

Dodecyltrimethylammonium chloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:112-00-5
:
Formula:C₁₅H₃₄ClN
MDL Number:MFCD00041974
:263.89 g/mol
Melting Pt:237 °C (dec.)
Storage Temperature:Ambient
EINECS:203-927-0
UN:3077
ADR:9,III
:GHS07
:GHS09
Specification Test Results
- Appearance:White to light yellow Powder and/or chunks
- Infrared spectrum:Conforms
- Titration Argentometric:≥97.5 %
- Solubility:(10 % in water) Clear
- Color of solution:≤100 APHA (10 % in water)
Specification Test Results
Frequently Bought Together