
Tetradecyltrimethylammonium bromide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1119-97-7
:8044-71-1
Formula:C₁₇H₃₈BrN
MDL Number:MFCD00011770
:336.394 g/mol
Merck Index:14,6336
Melting Pt:245…250 °C
Storage Temperature:Ambient
EINECS:214-291-9
UN:1759
ADR:8,III
:GHS05
Related Information
Frequently Bought Together