Dodecyltrimethylammonium bromide, Supelco®
Catalog # 44239-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:308.35 g/mol
Melting Pt:264…266 °C
:M-Clarity™ quality level = MQ100
EINECS:214-290-3
CAS Number:1119-94-4
Formula:C₁₅H₃₄BrN
MDL Number:MFCD00011767