
1,2-Bis(diphenylphosphino)ethane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1663-45-2
:
Formula:C₂₆H₂₄P₂
MDL Number:MFCD00003047
:398.42 g/mol
Melting Pt:140…142 °C
:M-Clarity™ quality level = MQ200
EINECS:216-769-2
:GHS07
Storage Temperature:Ambient
Frequently Bought Together