Bismuth(III) nitrate basic, Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
Synonyms: Bismuth(III) nitrate alkaline, Bismuth hydroxide nitrate oxide, Bismuth(III) hydroxide nitrate oxide, Bismuth subnitrate
- CAS Number:1304-85-4
- Formula:Bi₅O(OH)₉(NO₃)₄
- MDL Number:MFCD00064844
- Molecular Weight:1461.99 g/mol
- Density:4.928 g/cm³ (20 °C)
- EINECS:215-136-8
- UN:1477
- ADR:5.1,II
- :M-Clarity™ quality level = MQ200
2 Options Available Below
Product Details & Documents
Synonyms: Bismuth(III) nitrate alkaline, Bismuth hydroxide nitrate oxide, Bismuth(III) hydroxide nitrate oxide, Bismuth subnitrate
- CAS Number:1304-85-4
- Formula:Bi₅O(OH)₉(NO₃)₄
- MDL Number:MFCD00064844
- Molecular Weight:1461.99 g/mol
- Density:4.928 g/cm³ (20 °C)
- EINECS:215-136-8
- UN:1477
- ADR:5.1,II
- :M-Clarity™ quality level = MQ200
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



