Order Entry
Czech Republic
ContactUsLinkComponent
tert-Butylchlorodimethylsilane for synthesis, Sigma-Aldrich®
Flammable
Corrosive
tert-Butylchlorodimethylsilane for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Corrosive
tert-Butylchlorodimethylsilane for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) (decomposition)
  • Melting Point:
    87 - 90 °C
  • Molar Mass:
    150.72 g/mol
  • Boiling Point:
    125 °C
  • Flash Point:
    22 °C
  • Synonyms:
    tert-Butyldimethylsilyl chloride, tert-Butylchlorodimethylsilane, TBDMSCl
  • Hill Formula:
    C6H15ClSi
  • Chemical Formula:
    (CH3)3CSi(CH3)2Cl

Specification Test Results

Related Information

Frequently Bought Together