

tert-Butylchlorodimethylsilane ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:18162-48-6
:
Formula:C₆H₁₅ClSi
MDL Number:MFCD00000501
:150.72 g/mol
Boiling Pt:124…126 °C
Melting Pt:86…89 °C
Density:0.81
Flash Pt:22 °C (72 °F)
Storage Temperature:Ambient
EINECS:242-042-4
ADR:4.1,II
:GHS02
:GHS05
Related Information
Frequently Bought Together