Quartz, fine-granular analytical reagent, washed, calcinated, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:14808-60-7
:7631-86-9
Formula:SiO₂
MDL Number:MFCD00217788
:60.08 g/mol
Boiling Pt:2230 °C (1013 hPa)
Melting Pt:1427 °C
Density:2.622…2.645 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:238-878-4
Related Information
Frequently Bought Together