Order Entry
Czech Republic
ContactUsLinkComponent
 
Boron nitride ≥99.5% (metals basis), rods low binder, Length: 203mm, Ø 9.5mm
Katalogové číslo 45575.KG
CAS Number:  
Boron nitride ≥99.5% (metals basis), rods low binder, Length: 203mm, Ø 9.5mm
Katalogové číslo 45575.KG
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :24.82 g/mol
Merck Index:14,1346
Melting Pt:2527 °C
Density:2.15 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-136-6
CAS Number:10043-11-5
Formula:BN
MDL Number:MFCD00011317