Potassium dihydrogen phosphate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7778-77-0
:
Formula:KH₂PO₄
MDL Number:MFCD00011401
:136.09 g/mol
Merck Index:13,07744
Boiling Pt:∼100 °C (1013 hPa)
Melting Pt:253 °C
Density:2.3 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:231-913-4
REACH:01-2119490224-41
Frequently Bought Together