Order Entry
Czech Republic
ContactUsLinkComponent
Potassium hydroxide, pellets NF, Multi-Compendial, J.T.Baker®
Corrosive
Irritant
Potassium hydroxide, pellets NF, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Corrosive
Irritant
Potassium hydroxide, pellets NF, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Bulk Pharmaceutical Chemical:
    Passes Test
  • NF - Identification A:
    Passes Test
  • NF - Identification B:
    Passes Test
  • NF - Insoluble Substances:
    Passes Test
  • NF - Content of Potassium:
    ≥59.9 %
  • NF - Limit of Sodium:
    ≤1.0 %
  • NF - Assay (Total Alkali):
    ≥85.0 %
  • NF - Carbonate (as K₂CO₃):
    ≤3.5 %
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Carbonate (as K₂CO₃):
    ≤2.0 %
  • EP/BP - Chloride (Cl):
    ≤200 ppm
  • EP/BP - Phosphate (PO₄):
    ≤100 ppm
  • EP/BP - Sulfate (SO₄):
    ≤200 ppm
  • EP/BP - Iron (Fe):
    ≤10 ppm
  • EP/BP - Sodium (Na):
    ≤1.0 %
  • EP/BP - Assay:
    85.0 - 100.5 %
  • JP - Identification A:
    Passes Test
  • JP - Identification B:
    Passes Test
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Chloride (Cl):
    ≤0.050 %
  • JP - Heavy Metals (as Pb):
    ≤30 ppm
  • JP - Sodium (Na):
    Passes Test
  • JP - Potassium Carbonate (K₂CO₃):
    ≤2.0 %
  • JP - Assay (KOH):
    ≥85.0 %
  • No Class 1,2,3 or other solvents are used or produced in the:
    = 85.0 %

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together