1-(4-Methoxyphenyl)piperazine, Sigma-Aldrich®
Catalog # SIAL571415-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:192.26 g/mol
Boiling Pt:118 °C (0.3 mmHg)
Melting Pt:261…262 °C
:M-Clarity™ quality level = MQ100
EINECS:253-829-7
CAS Number:38212-30-5
Formula:C₁₁H₁₆N₂O
MDL Number:MFCD00040736