
1-(2-Hydroxyethyl)piperazine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:103-76-4
:
Formula:C₆H₁₄N₂O
MDL Number:MFCD00005970
:130.19 g/mol
Boiling Pt:242…246 °C
Melting Pt:–39 °C
Density:1.060
Flash Pt:135 °C (275 °F)
EINECS:203-142-3
:GHS05
Storage Temperature:Ambient
Frequently Bought Together