
Cyclohexanone oxime ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:100-64-1
:
Formula:C₆H₁₁NO
MDL Number:MFCD00001660
:113.16 g/mol
Boiling Pt:206…210 °C
Melting Pt:87…91 °C
Density:1.13 g/cm³ (20 °C)
Flash Pt:103 °C (217 °F)
EINECS:202-874-0
:GHS06
Storage Temperature:Ambient
Related Information
Frequently Bought Together