

Tetrachloro-p-benzoquinone, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:118-75-2
:
Formula:C₆Cl₄O₂
MDL Number:MFCD00001594
:245.88 g/mol
Merck Index:13,02088
Boiling Pt:270 °C (1013 hPa)
Melting Pt:290…293 °C
Density:1.97 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:204-274-4
:GHS07
:GHS09
Frequently Bought Together