Order Entry
Czech Republic
ContactUsLinkComponent
Tetrachloro-p-benzoquinone 99%
Tetrachloro-p-benzoquinone 99%
Certificates
undefined
Tetrachloro-p-benzoquinone 99%

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Yellow to yellow-green Powder or crystals
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.5 % (1 g, 105°C, 3 hrs)
  • HPLC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together