


Chloramine-T (sodium salt) trihydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7080-50-4
:
Formula:CH₃C₆H₄SO₂NClNa·3H₂O
MDL Number:MFCD00149066
Merck Index:13,02085
Melting Pt:167…170 °C
Density:1.43 g/cm³ (20 °C)
Flash Pt:192 °C (closed cup)
Storage Temperature:Ambient
EINECS:204-854-7
UN:3263
ADR:8,III
:M-Clarity™ quality level = MQ200
:281.69 g/mol
:GHS05
:GHS08
:GHS07
Frequently Bought Together