1,1-Dimethoxy-2-phenylethane, Sigma-Aldrich®
Catalog # SIALP16605-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:166.22 g/mol
Boiling Pt:221 °C (1013 hPa)
Melting Pt:62…64 °C
Density:1 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:202-945-6
CAS Number:101-48-4
Formula:C₆H₅CH₂CH(OCH₃)₂
MDL Number:MFCD00008487