
4'-Fluoro-2'-hydroxyacetophenone ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1481-27-2
:
Formula:C₈H₇FO₂
MDL Number:MFCD00143203
Boiling Pt:65 °C (2 mmHg)
Melting Pt:29…34 °C
Flash Pt:98 °C (208 °F)
:154.14 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together