5'-Fluoro-2'-hydroxyacetophenone, Sigma-Aldrich®
Catalog # SIAL247170-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:154.14 g/mol
Boiling Pt:182 °C (1013 hPa)
Melting Pt:56 °C
:M-Clarity™ quality level = MQ200
EINECS:206-893-5
CAS Number:394-32-1
Formula:C₈H₇FO₂
MDL Number:MFCD00011668