
4'-Hydroxy-3'-methoxyacetophenone 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:498-02-2
:
Formula:C₉H₁₀O₃
MDL Number:MFCD00008747
:166.18 g/mol
Merck Index:13,00746
Boiling Pt:295…300 °C (760 mmHg)
Melting Pt:112…116 °C
Storage Temperature:Ambient
EINECS:207-854-5
:GHS07
Specification Test Results
- Appearance:Yellow to brown Powder
- Infrared spectrum:Conforms
- Melting point:112 to 116 °C
- Water:≤0.5 % (K.F.)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together