2'-Hydroxy-6'-methoxyacetophenone, Sigma-Aldrich®
Catalog # SIAL303046-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:166.18 g/mol
EINECS:211-872-9
CAS Number:703-23-1
Formula:C₉H₁₀O₃
MDL Number:MFCD00008732