
2'-Hydroxy-4'-methoxyacetophenone ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:552-41-0
:
Formula:C₉H₁₀O₃
MDL Number:MFCD00008730
:166.18 g/mol
Boiling Pt:154 °C (20 mmHg)
Melting Pt:48…50 °C
Density:1.1604 g/cm³ (40 °C)
Flash Pt:>110 °C (230 °F)
EINECS:209-012-2
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together