Order Entry
Czech Republic
ContactUsLinkComponent
2',6'-Dichloro-3'-fluoroacetophenone 97%
Irritant
2',6'-Dichloro-3'-fluoroacetophenone 97%
Catalog # ACRO345511000
CAS Number:  
undefined
Irritant
2',6'-Dichloro-3'-fluoroacetophenone 97%
Catalog # ACRO345511000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5275 to 1.5295 (20°C, 589 nm)
  • GC:
    ≥96.0 %

Specification Test Results

About this item

 :
 :207.03 g/mol
Flash Pt:110 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:290835-85-7
Formula:C₈H₅Cl₂FO
MDL Number:MFCD02093760