Methyl-3,4,5-trihydroxybenzoate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:99-24-1
:
Formula:C₈H₈O₅
MDL Number:MFCD00002194
Merck Index:13,04366
Boiling Pt:450 °C (1013 hPa)
Melting Pt:200…203 °C
:M-Clarity™ quality level = MQ100
EINECS:202-741-7
Frequently Bought Together