
Isophthaloyl dichloride for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:99-63-8
:
Formula:C₈H₄Cl₂O₂
MDL Number:MFCD00000678
:203.02 g/mol
Boiling Pt:276 °C (1013 hPa)
Melting Pt:42…45 °C
Density:1.388 g/cm³ (17 °C)
EINECS:202-774-7
ADR:8,III
:M-Clarity™ quality level = MQ200
UN:3261
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) (decomposition)
- Melting Point:42 - 45 °C
- Molar Mass:203.03 g/mol
- Boiling Point:276 °C
- Vapor Pressure:0.04 hPa (25 °C)
- Flash Point:180 °C
- Explosion Limit:1.5 - 8.9 %(V)
- Density:1.39 g/cm3 (17 °C)
Specification Test Results
Frequently Bought Together