4-Chloro-2-methoxyphenol
Catalog # EHERC11415800
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
- Environmentally Preferable:
Specifications
About this item
Synonyms: 5-Chlor-2-hydroxyanisole, 4-Chloroguaiacol
:
:158.58 g/mol
CAS Number:16766-30-6
Formula:C₇H₇ClO₂
MDL Number:MFCD00070774
Boiling Pt:241
Density:1.304 g/cm³ (20 °C)