
1-(p-Tolyl)ethylamine ≥96.0% (by GC, titration analysis)
TCIAT2505-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 96.0 %
- Purity (Nonaqueous Titration):min. 96.0 %
:
:135.21 g/mol
:204°C
:81°C
:443-160-0
:2735
:8,III
:GHS05
:586-70-9
:C₉H₁₃N
:MFCD02177111