
(R)-(-)-2,2,2-Trifluoro-1-(9-anthryl)ethanol ≥99.0% (by GC) for e.e. determination by NMR
Catalog # TCIAT1520-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Light yellow to Green powder to crystal
- Purity (GC):min. 99.0 %
- Optical purity (GC):min. 99.0 ee%
- Melting point:134.0 to 136.0 deg-C
Specification Test Results
About this item
:
:276.26 g/mol
EINECS:258-611-5
:GHS07
CAS Number:53531-34-3
Formula:C₁₆H₁₁F₃O
MDL Number:MFCD00001260