Order Entry
Czech Republic
ContactUsLinkComponent
Sulphonyl chloride polystyrene resin 4.5 - 5.3 mmol/g, cross-linked with 1% DVB (50-100 mesh)
Corrosive
Sulphonyl chloride polystyrene resin 4.5 - 5.3 mmol/g, cross-linked with 1% DVB (50-100 mesh)
Catalog # TCIAS0545-5G
Supplier:  TCI
CAS Number:  
Corrosive
Sulphonyl chloride polystyrene resin 4.5 - 5.3 mmol/g, cross-linked with 1% DVB (50-100 mesh)
Catalog # TCIAS0545-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Orange to Brown to Dark red powder to crystal
  • Loading:
    4.5 to 5.3 mmol/g

Specification Test Results

About this item

 :
UN:3261
ADR:8,III
 :GHS05
CAS Number:163894-16-4
MDL Number:MFCD00211665