Phenylthiohydantoinglutamic acid ≥95.0% (by HPLC, titration analysis)
Catalog # TCIAP0375-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Neutralization titration):min. 95.0 %
- Melting point:172.0 to 177.0 deg-C
Specification Test Results
About this item
:
:264.30 g/mol
EINECS:227-056-0
CAS Number:5624-27-1
Formula:C₁₂H₁₂N₂O₃S
MDL Number:MFCD00070137