Order Entry
Czech Republic
ContactUsLinkComponent
Phenylhydrazine sulphate ≥98.0% (by titrimetric analysis)
Toxic
Health
Environment
Phenylhydrazine sulphate ≥98.0% (by titrimetric analysis)
Catalog # TCIAP0186-25G
Supplier:  TCI
CAS Number:  
Toxic
Health
Environment
Phenylhydrazine sulphate ≥98.0% (by titrimetric analysis)
Catalog # TCIAP0186-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (Potassium iodate Method):
    min. 98.0 %

Specification Test Results

About this item

 :
 :314.36 g/mol
EINECS:257-622-2
UN:2811
ADR:6.1,III
 :GHS06
 :GHS08
 :GHS09
CAS Number:52033-74-6
Formula:C₆H₈N₂
MDL Number:MFCD00070608