α-Linolenic acid methyl ester, Sigma-Aldrich®
Catalog # SIAL62210-10ML-F
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- Seq:2275
Specifications
About this item
:
:292.46 g/mol
Boiling Pt:364 °C (1013 hPa)
Melting Pt:–57 °C
Density:0.8982 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:206-102-3
CAS Number:301-00-8
Formula:C₁₉H₃₂O₂
MDL Number:MFCD00135851