Zadání objednávky
Czech Republic
ContactUsLinkComponent
Toluen-α-sulfonylfluorid
Toxic
Corrosive
Toluen-α-sulfonylfluorid
Catalog # ROTH6367.1
Supplier:  Roth Carl
CAS Number:  
Toxic
Corrosive
Toluen-α-sulfonylfluorid
Catalog # ROTH6367.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Typ balení:
    Glass bottle for solids
  • Bal.:
    5 g
  • Sekvence:
    3173

Specifications

About this item

 :
Boiling Pt:80…85 °C (0,3 mmHg)
Melting Pt:91…92 °C
Storage Temperature:Ambient
EINECS:206-350-2
UN:2923
ADR:8,III
 :GHS06
 :GHS05
CAS Number:329-98-6
Formula:C₇H₇FO₂S
MDL Number:MFCD00007424