2-Methyl-1-(p-tolyl)-2-propanol ≥98.0% (by GC)
Catalog # TCIAM2019-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:40.0 to 44.0 deg-C
Specification Test Results
About this item
:
:164.25 g/mol
Boiling Pt:112°C
EINECS:244-072-3
CAS Number:20834-59-7
:MFCD00021809
Formula:C₁₁H₁₆O
MDL Number:MFCD00029997