Order Entry
Czech Republic
ContactUsLinkComponent
(-)-2,2'-Methylenebis[(3aS,8aR)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole] ≥98.0% (by HPLC, total nitrogen)
Irritant
(-)-2,2'-Methylenebis[(3aS,8aR)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole] ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAM1402-500MG
Supplier:  TCI
CAS Number:  
Irritant
(-)-2,2'-Methylenebis[(3aS,8aR)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole] ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAM1402-500MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    500 mg
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Optical purity (LC):
    min. 98.0 ee%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    216.0 to 224.0 deg-C

Specification Test Results

About this item

 :
 :330.39 g/mol
 :GHS07
CAS Number:175166-49-1
Formula:C₂₁H₁₈N₂O₂
MDL Number:MFCD00799561