Order Entry
Czech Republic
ContactUsLinkComponent
3,3,5-Trimethylcyclohexyl mandelate (mixture of isomers) ≥94.0% (by GC)
3,3,5-Trimethylcyclohexyl mandelate (mixture of isomers) ≥94.0% (by GC)
Catalog # TCIAM0821-25G
Supplier:  TCI
CAS Number:  
3,3,5-Trimethylcyclohexyl mandelate (mixture of isomers) ≥94.0% (by GC)
Catalog # TCIAM0821-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 94.0 % (total of isomers)

Specification Test Results

About this item

 :
Merck Index:14,02704
 :276.38 g/mol
EINECS:207-271-6
CAS Number:456-59-7
Formula:C₁₇H₂₄O₃
MDL Number:MFCD00056623