Methylfluorone Hemisulphate for detection of Sb, Ge, Mo, Sn
Catalog # TCIAM0678-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
Specification Test Results
About this item
:
:307.27 g/mol
Formula:C₁₄H₁₀O₅
MDL Number:MFCD00022274