
N-(2-Hydroxypropyl)morpholine ≥98.0% (by GC)
Catalog # TCIAH0437-25ML
Supplier: TCI
CAS Number:
Specifications
- Pk:25 mL
- Seq:2315
Specifications
Specification Test Results
- Appearance:Colorless to Yellow to Orange clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
About this item
:
Boiling Pt:218°C
Flash Pt:92°C
:145.20 g/mol
EINECS:218-298-8
:GHS07
CAS Number:2109-66-2
Formula:C₇H₁₅NO₂
MDL Number:MFCD00023378