

Thorin hydrate ≥98.0% (by HPLC, titration analysis) for determination of Th, etc.
Catalog # TCIAH0240-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Orange to Brown to Dark red powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Iodometric Titration):min. 98.0 % (after drying)
- Lambda max.:480.0 to 490.0 nm (H2O)
- Absorbance:min. 0.500 (10mg/L, H2O, 480.0 to 490.0 nm) (calcd.on dried substance)
Specification Test Results
About this item
:
:576.29 g/mol
EINECS:222-993-1
UN:3465
ADR:6.1,II
:GHS06
:GHS09
CAS Number:3688-92-4
Formula:C₁₆H₁₃AsN₂O₁₀S₂
MDL Number:MFCD00003888