
1,1,3,3-Tetraethoxypropane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:122-31-6
:
Formula:C₁₁H₂₄O₄
MDL Number:MFCD00009240
:220.31 g/mol
Boiling Pt:220 °C (1013 hPa)
Melting Pt:–90 °C
Density:0,9185 g/cm³ (20 °C)
Flash Pt:87 °C
EINECS:204-533-1
:GHS07
Storage Temperature:Ambient
Frequently Bought Together