Order Entry
Czech Republic
ContactUsLinkComponent
Ethanolamine ≥99.0% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Ethanolamine ≥99.0% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 9314-01
Supplier:  Avantor
CAS Number:  
undefined
Ethanolamine ≥99.0% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 9314-01
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, amber, narrow mouth
  • Pk:
    500 mL
  • Seq:
    2725
  • Environmentally Preferable:

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 99.0 %
  • Assay (HOCH₂CH₂NH₂) (by GC, corrected for water):
    ≥ 99.0 %
  • Color (APHA):
    ≤ 15
  • Water (H₂O):
    ≤ 0.30 %
  • Heavy Metals (by ICP-OES):
    ≤ 5 ppm
  • Trace Impurities - Iron (Fe):
    ≤ 5 ppm

Specification Test Results

About this item

Synonyms: 2-Aminoethanol, Monoethanolamine
 :
 :61.08 g/mol
Melting Pt:10.5
EINECS:205-483-3
UN:2491
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:141-43-5
Formula:NH₂CH₂CH₂OH
MDL Number:MFCD00008183
Boiling Pt:171
Density:1.0164 g/cm³ (20 °C)
Flash Pt:93
ADR:8 III
Merck Index:13,03762
 :Management of Change (MOC) category = N