
N,N-Dibenzylhydrazine ≥96.0% (by GC, titration analysis)
TCIAD2132-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:White to Orange to Green powder to crystaline
- Purity (GC):min. 96.0 %
- Purity (Nonaqueous Titration):min. 96.0 %
- Melting point:55.0 to 58.0 deg-C
- Solubility in Methanol:almost transparency
:
:212.30 g/mol
:171°C
:GHS07
:Refrigerator
:5802-60-8
:C₁₄H₁₆N₂
:MFCD00144976