Order Entry
Czech Republic
ContactUsLinkComponent
3-(Dimethylamino)-1,2-propanediol ≥98.0% (by GC, titration analysis)
Corrosive
3-(Dimethylamino)-1,2-propanediol ≥98.0% (by GC, titration analysis)
Catalog # TCIAD2072-25G
Supplier:  TCI
CAS Number:  
Corrosive
3-(Dimethylamino)-1,2-propanediol ≥98.0% (by GC, titration analysis)
Catalog # TCIAD2072-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %

Specification Test Results

About this item

 :
Boiling Pt:217°C
Flash Pt:105°C
UN:2735
ADR:8,III
 :119.1622 g/mol
EINECS:210-802-4
 :GHS05
CAS Number:623-57-4
Formula:C5H13NO2
MDL Number:MFCD00004714