
Dipropylamine hydrochloride ≥99.0% (by titrimetric analysis)
Catalog # TCIAD0931-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
About this item
:
:137.65 g/mol
:GHS07
CAS Number:5326-84-1
Formula:C₆H₁₅N
MDL Number:MFCD00054288