
Diphenylthiocarbazide ≥98.0% (by HPLC)
Catalog # TCIAD0917-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
Specification Test Results
About this item
:
:258.34 g/mol
EINECS:210-717-2
:GHS07
Storage Temperature:Refrigerator
CAS Number:622-03-7
Formula:C₁₃H₁₄N₄S
MDL Number:MFCD00046042