Order Entry
Czech Republic
ContactUsLinkComponent
2-Dimethylaminomethylphenol ≥70% (by GC) (contains Phenol)
Corrosive
Irritant
Health
2-Dimethylaminomethylphenol ≥70% (by GC) (contains Phenol)
Catalog # TCIAD0655-25ML
Supplier:  TCI
CAS Number:  
Corrosive
Irritant
Health
2-Dimethylaminomethylphenol ≥70% (by GC) (contains Phenol)
Catalog # TCIAD0655-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Red to Green clear liquid
  • Purity (GC):
    min. 70.0 %

Specification Test Results

About this item

 :
 :151.21 g/mol
EINECS:204-413-9
UN:1760
ADR:8,II
 :GHS05
 :GHS07
 :GHS08
CAS Number:120-65-0
Formula:C₉H₁₃NO
MDL Number:MFCD00045793